missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BHLHE40 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17895-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a member of the nucleoporin family. The protein is localized to the nuclear rim and is an essential component of the nuclear pore complex (NPC). All molecules entering or leaving the nucleus either diffuse through or are actively transported by the NPC. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.Specifica
BHLHE40 | |
Polyclonal | |
Unconjugated | |
BHLHE40 | |
BHLHB2, BHLHE40, DEC1, SHARP 2, SHARP2, STRA13, Stra14 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20893, 79431, 8553 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14503, O35185, O35780 | |
BHLHE40 | |
BHLHE40 Fusion Protein Ag12236 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |