Learn More
MP Biomedicals™ BigPrep™ Lysing Matrix Z, 50 mL Tubes
BigPrep™ Lysing Matrix Z contains 2.0 mm diameter yttria-stabilized zirconium oxide beads and is used to grind soft animal and plant tissues and insects.
Marca: MP Biomedicals™ 116979050
596.83 EUR valido fino al 2024-12-31
Usa il codice promo "23257" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 2.49400kg
Descrizione
Lysing Matrix Z tubes can be used with FastPrep instruments or similar bead beaters. They are ideal for lysis of animal and plant tissue and whole insects. Lysing Matrix Z is an alternative to the very aggressive Lysing Matrix A for applications such as RNA and protein isolation from very tough tissue.
- Tube size: 50 mL
- Samples types: Animal tissue, plant tissue, tough stems, roots, seeds, whole insects
- Matrix composition: 2 mm diameter yttria-stabilized zirconium oxide beads
Specifica
Yttria-stabilized Zirconium Oxide | |
Animal Tissues, Plants, Insects | |
FastPrep instruments or other bead beaters | |
Lysing Matrix Z | |
50 x 50 mL |
Room temperature | |
Dnase-free | |
Nucleic Acid Isolation and Purification, Homogenization | |
Lysis Matrix Tube | |
Rnase-free |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.