missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Cynomolgus CD24 Protein, Fc Tag, ultra sensitivity (primary amine labeling)

Marca:  ACROBiosystems CD4-C8254-200ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 17246100

  • 2011.89€ / 200µg
Spedizione stimata: 13-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

CD24 may have a pivotal role in cell differentiation of different cell types. Signaling could be triggered by the binding of a lectin-like ligand to the CD24 carbohydrates, and transduced by the release of second messengers derived from the GPI-anchor. Modulates B-cell activation responses. Promotes AG-dependent proliferation of B-cells, and prevents their terminal differentiation into antibody-forming cells. In association with SIGLEC10 may be involved in the selective suppression of the immune response to danger-associated molecular patterns (DAMPs) such as HMGB1, HSP70 and HSP90. Plays a role in the control of autoimmunity.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 29.3 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifica

Specifica

29.3 kDa
Biotinylated Cynomolgus CD24, Fc Tag, primary amine labeling (CD4-C8254) is expressed from human 293 cells (HEK293). It contains AA Thr 27 - Lys 54 (Accession # I7GKK1-1).
-20°C
CD24
Cynomolgus Monkey
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins
200 μg
Thr 27 - Lys 54
CD24,CD24A
UltraLys
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE.
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali