Learn More
ACROBiosystems Biotinylated Human Syndecan-1 Protein, Fc, Avitag™
Biotinylated Protein
Marca: ACROBiosystems SY1-H82F3-25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Neuronal protein that plays several roles in synaptic activity such as regulation of synaptic vesicle trafficking and subsequent neurotransmitter release. Participates as a monomer in synaptic vesicle exocytosis by enhancing vesicle priming, fusion and dilation of exocytotic fusion pores. Mechanistically, acts by increasing local Ca2+ release from microdomains which is essential for the enhancement of ATP-induced exocytosis. Acts also as a molecular chaperone in its multimeric membrane-bound state, assisting in the folding of synaptic fusion components called SNAREs (Soluble NSF Attachment Protein REceptors) at presynaptic plasma membrane in conjunction with cysteine string protein-alpha/DNAJC5. This chaperone activity is important to sustain normal SNARE-complex assembly during aging. Plays also a role in the regulation of the dopamine neurotransmission by associating with the dopamine transporter (DAT1) and thereby modulating its activity.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 52.0 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.Specifica
52.0 kDa | |
Biotinylated Human Syndecan-1, Fc,Avitag (SY1-H82F3) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Gly 254 (Accession # P18827-1). | |
-20°C | |
Syndecan-1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
25 μg | |
Gln 23 - Gly 254 | |
SDC1,Syndecan-1,CD138,SYND1,SDC | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |