missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLVRB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17727-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The final step in heme metabolism in mammals is catalyzed by the cytosolic biliverdin reductase enzymes A and B (EC 1.3.1.24).Specifica
BLVRB | |
Polyclonal | |
Unconjugated | |
BLVRB | |
Biliverdin IX beta reductase, Biliverdin reductase B, BLVRB, BVR B, BVRB, Flavin reductase, FLR, FR, GHBP, Green heme binding protein, NADPH dependent diaphorase, NADPH flavin reductase, SDR43U1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
233016, 292737, 645 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P30043, Q923D2 | |
Blvrb | |
BLVRB Fusion Protein Ag12124 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |