Learn More
Robu™ Imbuti filtranti in vetro borosilicato Buchner
Marca: Robu™ 21552
Dettagli aggiuntivi : Peso : 0.10000kg
Specifications
100 mm (OD); 95mm (ID) | |
22 mm | |
100 mm | |
100mm | |
Imbuto per filtrazione Buchner |
500 ml | |
Vetro borosilicato | |
Highly resistant to most chemicals such as solvents, acids and alkalis | |
2 |