Learn More
Lenz Laborglasinstrumente™ Bottiglia per lavaggio gas in vetro borosilicato con disco filtrante
Provides an inexpensive but effective method for the washing and drying of gases. Lenz Laborglasinstrumente™ Gas Washing Bottles Acc. to Drechsel with Filter Disc are manufactured from durable DURAN™ borosilicate glass.
Marca: Lenz Laborglasinstrumente™ 05510158
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Acc. to DIN 12596, complete with head
- Corrosion-free and resistant to thermal shock
- Available in a range of capacities
Specifica
500 ml | |
Flacone per lavaggio gas con disco filtrante | |
1 | |
DIN 12596 |
Vetro borosilicato | |
Disco filtrante | |
NS 29/32 | |
Confezione da 1 |