Learn More
BPTF Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 28016-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00020kg
Codice prodotto. 17241396
  • 399.00€ / 150µL
Spedizione stimata: 15-05-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Bromodomain PHD finger transcription factor (BPTF), a histone-binding component of NURF (nucleosome-remodeling factor), plays an important role in catalyzing ATP-dependent nucleosome sliding and facilitating transcription of chromatin. It has been reported that BPTF maybe a novel and potential therapeutic target in hepatocellular carcinoma. BPTF may also regulate transcription through direct binding to DNA or transcription factors.(PMID: 30419422)
Specifica
Specifica
BPTF
Polyclonal
Unconjugated
150 μL
2186
Liquid
Immunohistochemistry, ELISA
300 μg/mL