Learn More
BPTF Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 28016-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00020kg
Descrizione
Bromodomain PHD finger transcription factor (BPTF), a histone-binding component of NURF (nucleosome-remodeling factor), plays an important role in catalyzing ATP-dependent nucleosome sliding and facilitating transcription of chromatin. It has been reported that BPTF maybe a novel and potential therapeutic target in hepatocellular carcinoma. BPTF may also regulate transcription through direct binding to DNA or transcription factors.(PMID: 30419422)
Specifica
BPTF | |
Polyclonal | |
Unconjugated | |
150 μL | |
2186 | |
Liquid |
Immunohistochemistry, ELISA | |
300 μg/mL | |
Rabbit | |
Primary | |
Human |