Learn More
Aalborg™ Valvola standard in ottone
Aalborg™ Precision Valve for Brass Flowmeter is designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
Brand: Aalborg™ VM5-BB
Additional Details : Peso : 0.80000kg
Description
- For high precision
- Designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
- They may also be used as shut off valves.
- Bodies of the valves are reinforced by structurally rigid metallic shells.
- Shells are made of brass, bushings are made of plated brass or 316 stainless steel.
Specifications
200 ml/min.;;en; | |
6200 ml/min.;;en; | |
Per flussometro gas/liquido da 150 mm | |
Valvola standard |