Learn More
Gilac™ Bread Container
Bread Container stacks when empty; perforated for increased product conservation, quality, and freshness.
Marca:  Gilac™ G180423
Dettagli aggiuntivi : Peso : 5.34000kg
Codice prodotto. 17918861
  • 131.60€ / Pezzo
Spedizione stimata: 12-06-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • Bread container, stacking when empty.
  • Perforated for increased product conservation quality and freshness.
  • 150 L: holds 55/60 250 g baguettes or 40/45 400 g loaves.
  • Compatible with GILAC 4 and 6-wheel dollies.
Specifica
Specifica
Bread Container
Beige
180 L
Polyethylene
47.6 gal.