Learn More
BUCHI Palloni di raccolta per evaporatori rotanti
Use these receiving flasks with any rotary evaporator. BUCHI Spherical Receiving Flasks offer high chemical resistance against acids, alkalis and organic substances. Ideal for collecting condensed solvents.
Marca: BUCHI 000422
Dettagli aggiuntivi : Peso : 0.07000kg
Descrizione
Use these receiving flasks with any rotary evaporator. Buchi™ Spherical Receiving Flasks offer high chemical resistance against acids, alkalis and organic substances. Ideal for collecting condensed solvents.
- Con giunto sferico in vetro KS–35
- Range compreso tra 50 ml e 3000 ml
- In vetro o con rivestimento in plastica
Specifica
Vetro standard | |
Evaporatore rotante | |
Senza rivestimento | |
100 ml |
Fiasca di raccolta | |
Giunto sferico KS-35 | |
Rotondo |