Learn More
BUCHI Palloni di raccolta per evaporatori rotanti
Use these receiving flasks with any rotary evaporator. BUCHI Spherical Receiving Flasks offer high chemical resistance against acids, alkalis and organic substances. Ideal for collecting condensed solvents.
Marca: BUCHI 000425
Dettagli aggiuntivi : Peso : 0.24000kg
Descrizione
Use these receiving flasks with any rotary evaporator. Buchi™ Spherical Receiving Flasks offer high chemical resistance against acids, alkalis and organic substances. Ideal for collecting condensed solvents.
- Dispone di un giunto sferico in vetro KS–35
- Intervallo delle dimensioni da 50 ml a 3000 ml
- Disponibile in vetro o con rivestimento in plastica
Specifica
Vetro standard | |
Evaporatore rotante | |
Senza rivestimento | |
100 ml |
Fiasca di raccolta | |
Giunto sferico KS-35 | |
Rotondo |