Learn More
Fisherbrand™ Buffer Saver Blocks
Physically reduce volume in positive and negative compartments of gel chamber thus reducing buffer requirements.
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Economic low gel and buffer volumes
- Available for multiSUB Horizontal Gel Units
Specifica
Sub-Gel Midi Horizontal Gel Electrophoresis Unit | |
Buffer Saver Block | |
2 Each |