missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 7.00, NIST, Chem-Lab
For laboratory use, pH measurements, traceable to SRM from NIST
Marca: Chem Lab CL03.0216.9025
Dettagli aggiuntivi : Peso : 20.20000kg
Descrizione
pH = 7.13 at 0°CpH = 7.05 at 10°C
pH = 7.00 at 20°C
pH = 6.99 at 25°C
pH = 6.98 at 30°C
pH = 6.97 at 40°C
pH = 6.96 at 50°C
pH = 6.96 at 60°C
pH = 6.97 at 70°C
pH = 6.98 at 80°C
pH = 7.00 at 90°C
Specifica
Phosphate | |
7.00 | |
PE |
1.01g/mL | |
25 L | |
NIST traceable |