Learn More
Abeba™ Business Men 32922 Shoes
ESD dissipative with high wearing comfort. Smooth leather construction.
Marca: Abeba™ 32922/43
Dettagli aggiuntivi : Peso : 1.15000kg
Descrizione
- CE EN ISO 20347:2012 OB A SRA
- Smooth leather
- Black
- Slip resistance special rubber outsole sole
- ESD dissipative according to DIN EN 61340
- Lining microfiber, respiration absorbing
- Instep area with lacing
- Insole leather
Specifica
CE EN ISO 20347:2012 OB A SRA | |
Microfiber | |
43 | |
Rubber |
Black | |
Leather | |
Slip-resistant | |
32922 |