missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C19orf43 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19420-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The antibody was raised from the C19orf43 protein. A band at ∽28kd was detected in Western Blot with human placenta, 293 cell, and A431 cell lysates.
C19orf43, also named as MGC2803 and fSAP18, is a protein coding gene. Exoribonuclease that is part of the telomerase RNA 3′ end processing complex and which has the ability to all four unpaired RNA nucleotides from 5′ end or 3′ end with higher efficiency for purine bases.Specifica
C19orf43 | |
Polyclonal | |
Unconjugated | |
C19orf43 | |
C19orf43, fSAP18 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
288920, 68544, 79002 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BQ61 | |
C19orf43, Trir | |
C19orf43 Fusion Protein Ag13747 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |