missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sensing Receptor Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19125-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The 60-65 kDa band detected by this antibody might represent a degradation product of CASR.
Senses changes in the extracellular concentration of calcium ions. The activity of this receptor is mediated by a G- protein that activates a phosphatidylinositol-calcium second messenger system.Specifica
Calcium Sensing Receptor | |
Polyclonal | |
Unconjugated | |
CASR | |
Calcium Sensing Receptor, CAR, CASR, FHH, FIH, GPRC2A, HHC, HHC1, NSHPT, PCAR1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12374, 24247, 846 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P41180, P48442, Q9QY96 | |
CASR | |
Peptide | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |