missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR7 (CD197) Mouse anti-Human , APC, Clone: 2-L1-A, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566762
Dettagli aggiuntivi : Peso : 0.00060kg
Descrizione
CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow CytometrySpecifica
CCR7 (CD197) | |
Monoclonal | |
APC | |
BLR2; CC chemokine receptor 7; CMKBR7; EBI1; EVI1; Epstein-Barr virus induced gene 1; MIP-3 beta receptor | |
Human CCR7 Transfected Cell Line | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
2-L1-A | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to APC under optimum conditions, and unconjugated antibody and free APC were removed. |