missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD100 Mouse anti-Human, Clone: 1C1A6, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66582-1-IG-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Semaphorins are neuronal chemorepellants that direct pioneering neurons during nervous system development. CD100 is a novel semaphorin that induces B cells to aggregate and improves their viability in vitro. CD100 modifies CD40-CD40L B-cell signaling by augmenting B-cell aggregation and survival and down-regulating CD23 expression. CD72 is a lymphocyte receptor for the Class IV semaphorin CD100, which is involved in regulating B cell signaling.Specifica
CD100 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q92854 | |
SEMA4D | |
CD100 Fusion Protein Ag26461 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1C1A6 | |
Unconjugated | |
SEMA4D | |
A8, BB18, C9orf164, CD100, coll 4, GR3, M sema G, SEMA4D, SEMAJ, Semaphorin 4D | |
Mouse | |
Protein G | |
RUO | |
10507 | |
-20°C | |
Liquid |