missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD107a Mouse anti-Human , Alexa Fluor 488, Clone: H4A3, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 567007
Dettagli aggiuntivi : Peso : 0.00020kg
Descrizione
CD107a Monoclonal antibody specifically detects CD107a in Human samples. It is validated for Flow CytometrySpecifica
CD107a | |
Monoclonal | |
Alexa Fluor 488 | |
LAMP1; LAMP-1; LAMPA; LGP120 | |
Human Adult Adherent Peripheral Blood Cells | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
H4A3 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 488 under optimum conditions, and unreacted Alexa Fluor 488 was removed. |