Learn More
CD16 Mouse anti-Human, Unconjugated, Clone: 1E12E11, Proteintech
Mouse Monoclonal Antibody
Marca:  Proteintech 66779-1-IG-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17234646
  • 140.00€ / 20µL
Spedizione stimata: 20-05-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
CD16 is a 50-70-kDa low affinity Fc receptor found on the surface of natural killer cells, neutrophil polymorphonuclear leukocytes, monocytes and macrophages. CD16 mediates antibody-dependent cellular cytotoxicity (ADCC) and other antibody-dependent responses, such as phagocytosis. CD16 has been identified as Fc receptors FcγRIIIa (CD16a) and FcγRIIIb (CD16b), encoded by two nearly identical genes, FCGR3A and the FCGR3B.
Specifica
Specifica
CD16
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse
IgG1
Immunohistochemistry, ELISA
1E12E11
Unconjugated
20 μL
2214