missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Mouse anti-Human, Baboon, Cynomolgus, Rhesus, BUV805, Clone: 2H7, BD Horizon™
Mouse Monoclonal Antibody
Marca: BD Biosciences 612906
Dettagli aggiuntivi : Peso : 0.00020kg
Descrizione
CD20 Monoclonal antibody specifically detects CD20 in Human,Baboon,Cynomolgus,Rhesus samples. It is validated for Flow Cytometry.Specifica
CD20 | |
Monoclonal | |
Brilliant Ultraviolet 805 | |
25 Tests | |
Primary | |
Human, Baboon, Cynomolgus, Rhesus Monkey | |
IgG2b κ |
Flow Cytometry | |
2H7 | |
Mouse | |
RUO | |
CD20 | |
Refrigerate (2 to 8°C) |