missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200 Mouse anti-Human , APC, Clone: MRC OX-104, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566893
Dettagli aggiuntivi : Peso : 0.00060kg
Descrizione
CD200 Monoclonal antibody specifically detects CD200 in Human samples. It is validated for Flow CytometrySpecifica
CD200 | |
Monoclonal | |
APC | |
OX-2; MOX1; MOX2 ; My033 | |
Human CD200 Recombinant Protein | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
MRC OX-104 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to APC under optimum conditions, and unconjugated antibody and free APC were removed. |