missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD24 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18330-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. CD24 is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin′s lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation, and control of autoimmunity.Specifica
CD24 | |
Polyclonal | |
Unconjugated | |
CD24 | |
CD24, CD24 molecule, CD24A, Signal transducer CD24 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100133941, 12484 | |
-20°C | |
Liquid |
Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P24807, P25063 | |
CD24 | |
CD24 Fusion Protein Ag11679 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |