missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse anti-Human, Brilliant Ultraviolet 496, Clone: UCHT1, BD Horizon™
Mouse Monoclonal Antibody
Marca: BD Biosciences 612940
Dettagli aggiuntivi : Peso : 0.00060kg
Descrizione
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow cytometry.Specifica
CD3 | |
Monoclonal | |
Brilliant Ultraviolet 496 | |
CD3e; CD3E; T3E; TCRE; T-cell surface antigen T3/Leu-4 epsilon | |
Human infant thymocytes and peripheral blood lymphocytes from a Sézary Syndrome donor | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
UCHT1 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
BALB/c IgG1 κ |