missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Mouse anti-Human , APC-R700, Clone: OKT4, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566808
Dettagli aggiuntivi : Peso : 0.00060kg
Descrizione
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow CytometrySpecifica
CD4 | |
Monoclonal | |
APC-R700 | |
CD4 antigen (p55); T-cell surface antigen T4/Leu-3 | |
Sheep Erythrocyte Rosette-purified Human T Cells | |
RUO | |
Human | |
IgG2b κ |
Flow Cytometry | |
OKT4 | |
Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |