missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Mouse anti-Human, Brilliant Ultraviolet 661, Clone: SK3 (also known as Leu3a), BD Horizon™
Mouse Monoclonal Antibody
Marca: BD Biosciences 612962
Dettagli aggiuntivi : Peso : 0.00060kg
Descrizione
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow cytometry.Specifica
CD4 | |
Monoclonal | |
Brilliant Ultraviolet 661 | |
L3T4; Leu3a; T-cell surface antigen T4/Leu-3 ; W3/25 ; CD4 antigen (p55) | |
Human Peripheral Blood T Cells | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
SK3 (also known as Leu3a) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |