missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45.1 Mouse, PerCP-Cy5.5, Clone: A20, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio QAB42-PCP55-25ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
CD45.1 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Ptprc | |
Protein A/G Purified | |
RUO | |
19264 | |
IgG2a κ |
Flow Cytometry | |
A20 | |
PerCP-Cyanine5.5 | |
NP_001255215 | |
Mouse | |
25 μg | |
Primary | |
Store at 2-4°C. |