missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Mouse anti-Baboon, Human, Rhesus, , Brilliant Ultraviolet 737, Clone: SK1, BD Horizon™
Mouse Monoclonal Antibody
Marca: BD Biosciences 612754
Dettagli aggiuntivi : Peso : 0.00060kg
Descrizione
CD8 Monoclonal antibody specifically detects CD8 in Baboon,Human,Rhesus, samples. It is validated for Flow Cytometry.Specifica
CD8 | |
Monoclonal | |
Brilliant Ultraviolet 737 | |
Mouse | |
Affinity Purified | |
RUO | |
Baboon, Human, Rhesus Monkey | |
IgG1 κ |
Flow Cytometry | |
SK1 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human Peripheral Blood T Cells | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |