missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8a Mouse, V450, Clone: OKT8, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio QAB17-V450-100Tests
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
CD8a | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
CD8A | |
Protein A/G Purified | |
RUO | |
925 | |
IgG2a |
Flow Cytometry | |
OKT8 | |
Violet Fluor 450 (V450) | |
NP_741969 | |
Mouse | |
100 Tests | |
Primary | |
Store at 2-4°C. |