missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90.2 Rat, V450, Clone: 30-H12, enQuireBio™
Rat Monoclonal Antibody
Marca: EnquireBio QAB52-V450-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
CD90.2 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Thy1 | |
Protein A/G Purified | |
RUO | |
21838 | |
IgG2b |
Flow Cytometry | |
30-H12 | |
V450 | |
NP_033408 | |
Rat | |
100 μg | |
Primary | |
Store at 2-4°C. |