missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD95 (Fas) Mouse anti-Human, Baboon, Cynomolgus, Rhesus, BB700, Clone: DX2, BD Horizon™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566542
Dettagli aggiuntivi : Peso : 0.00060kg
Descrizione
CD95 (Fas) Monoclonal antibody specifically detects CD95 (Fas) in Human,Baboon,Cynomolgus,Rhesus samples. It is validated for Flow Cytometry.Specifica
CD95 (Fas) | |
Monoclonal | |
BB700 | |
100 Tests | |
Primary | |
Rhesus Monkey, Cynomolgus, Baboon, Human | |
IgG1 κ |
Flow Cytometry | |
DX2 | |
Mouse | |
RUO | |
CD95 (Fas) | |
Refrigerate (2 to 8°C) |