missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDR2L Mouse anti-Human, Mouse, Rat, Clone: 2E12D10, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66791-1-IG-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Cerebellar degeneration-related protein 2-like (CDR2L), also named as HUMPPA or Paraneoplastic 62 kDa antigen, is a 465 amino acid protein, which belongs to the CDR2 family. CDR2L contains three potential coiled-coil regions. The functions of protein is so far not known. CDR2L is expressed in the brain and localizes in the cytoplasm. The predicted MW of this protein is 53 kDa.Specifica
CDR2L | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A2A6T1, Q86X02 | |
CDR2L | |
CDR2L Fusion Protein Ag6139 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
2E12D10 | |
Unconjugated | |
CDR2L | |
CDR2L, HUMPPA, Paraneoplastic 62 kDa antigen | |
Mouse | |
Protein G | |
RUO | |
237988, 30850, 360656 | |
-20°C | |
Liquid |