Learn More
Cole-Parmer™ Check Valves with Pipe Thread Connections
Designed for use with Cole-Parmer tubing
Marca: Cole-Parmer™ 01350-26
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Check valve
- With pipe thread connections
- Available with 1/4 or 1/2 in. NPT(F) Connections
Specifica
Check Valve | |
1/4 in. NPT(F) | |
125 psi | |
81°C |
Polypropylene, Viton | |
1/4 in. NPT(F) | |
180°F |