Learn More
Cole-Parmer™ Check Valves with Pipe Thread Connections
Designed for use with Cole-Parmer tubing
Marca: Cole-Parmer™ 01350-24
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Check valve
- With pipe thread connections
- Available with 1/4 or 1/2 in. NPT(F) Connections
Specifica
Check Valve | |
1/2 in. NPT(F) | |
150 psi | |
60°C |
PVC, Viton | |
1/2 in. NPT(F) | |
140°F |