Learn More
WTW™ Chlorine Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
Marca: WTW™ 251402
Dettagli aggiuntivi : Peso : 0.80000kg
Specifica
DIN 38408 G4, EN ISO 7393 | |
5 mL | |
PhotoLab™ 6000 photometer, Photoflex™ meter | |
0.5 to 5.0 mg/L Cl2, free | |
Triazine |
3 to 4 at 20°C | |
Disinfectants, wastewater, drinking water | |
Iron | |
2°C to 8°C | |
Powder Pillows Kit |