Learn More
Macherey-Nagel™ Chromabond™ Alox B Polypropylene SPE Column
Marca: Macherey-Nagel™ 730429
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
3 mL | |
Solid Phase Extraction | |
Chromabond Alox B | |
150 m2/g |
SPE | |
60 to 150 μm | |
Solid Phase Extraction Column |