Learn More
Purolite™ Chromalite™ AD Monodispersed Polymeric Adsorbents for IEX and RP-HPLC
Chromalite™ AD monodispersed resins are hydrophobic resins made in monodisperse size for solid phase extraction, IEX-HPLC, RP-HPLC.
Marca:  Purolite™ LS06870-564
Dettagli aggiuntivi : Peso : 0.05000kg
Codice prodotto. 17089643
  • 654.00€ / 50g
Spedizione stimata: 04-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Manufactured using patented seeding technology foir uniform particle sizes
  • Solid Phase Extraction
  • Analytical Reverse Phase chromatography (RP-HPLC) and IEX chromatography (IEX-HPLC)
  • Adsorption of biomolecules
  • Extraction from biological fluid
  • High chemical stability
  • Efficient regeneration
  • Large surface area
  • Narrow particle size range
  • Excellent selectivity
  • Low extractable content
  • Compliant with FDA regulation
  • Compliant with ResAP(2004) 3
  • Halal, Kosher, TSE/BSE/GMO free
Specifica
Specifica
White to Off-White Spherical Beads
30AD2 Macroporous Polystyrene/Divinylbenzene Adsorbent
1 to 14 pH
>350 m2
Use within 5 years from date of manufacture
2°C to 20°C
24 to 36 μm
200 to 300 Å
50 g
20% ethanol slurry