Learn More
Chromalite™ M Methacrylic Hydrophilic Macroporous Resins, 40 to 90 μm, Purolite™
Chromalite™ M resins are hydrophilic, highly porous with high mechanical stability, based on a rigid methacrylic backbone. Optimal for protein purification and separation.
Marca: Chromalite LS04010-192
Dettagli aggiuntivi : Peso : 0.08000kg
Descrizione
Hydrophilic, macroporous resins based on a methacrylic backbone.
- Ion exchange chromatography
- Typical porosity of 1000Å suitable for protein purification (up to 250 kDa)
- Suitable for large-scale applications
- Excellent pressure-flow properties,designed for high performance, stability and reliability
- Purification of peptides obtained by fermentation
- High mechanical stability
- Uniform particle size
- Stable to CIP (up to 1M NaOH)
- Negligible swelling
- Compliant with ResAP(2004) 3
- Halal, Kosher and TSE/BSE/GMO free
Specifica
Wet | |
40 to 90 μm | |
100 mL | |
Epoxy |
Epoxy Methacrylate Macroporous Affinity Resin | |
1000 Å | |
Chromalite™ MEP/F |