Learn More
Purolite™ Chromalite™ PCG Macroporous Hydrophobic Polymeric Adsorbent
Chromalite™ PCG resins are hydrophobic porous polydivinylbenzene adsorbent. Porosity and matrix are specific for purification by reversed-phase chromatography, solid phase extraction and adsorption of biomolecules including proteins, peptides, oligonucleotides and antibiotics.
Marca:  Purolite™ LS06720-564
Dettagli aggiuntivi : Peso : 0.05000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17049553
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Descrizione
Descrizione
  • Reverse phase chromatography
  • Adsorption of biomolecules
  • Solid Phase Extraction
  • Highly hydrophobic
  • High chemical stability
  • More robust than silica materials
  • Easy column packing
  • Efficient regeneration
  • Compliant with FDA regulation
  • Compliant with ResAP(2004) 3
  • Halal, Kosher and TSE/BSE/GMO free
Specifica
Specifica
PCG1200F
2°C to 20°C
20 to 50 μm
300 to 500 Å
50 g
20% ethanol slurry
White to off-white spherical beads
Macroporous Polydivinylbenzene Adsorbent
1 to 14 pH
>600 m2/g
Use within 5 years from date of manufacture