Learn More
Purolite™ Chromalite™ PCG Macroporous Hydrophobic Polymeric Adsorbent
Chromalite™ PCG resins are hydrophobic porous polydivinylbenzene adsorbent. Porosity and matrix are specific for purification by reversed-phase chromatography, solid phase extraction and adsorption of biomolecules including proteins, peptides, oligonucleotides and antibiotics.
Marca: Purolite™ LS06730-192
Dettagli aggiuntivi : Peso : 0.25000kg
Descrizione
- Reverse phase chromatography
- Adsorption of biomolecules
- Solid Phase Extraction
- Highly hydrophobic
- High chemical stability
- More robust than silica materials
- Easy column packing
- Efficient regeneration
- Compliant with FDA regulation
- Compliant with ResAP(2004) 3
- Halal, Kosher and TSE/BSE/GMO free
Specifica
White to off-white spherical beads | |
2°C to 20°C | |
100 to 200 μm | |
300 to 500 Å | |
250 g | |
Wet |
PCG1200C | |
Macroporous Polydivinylbenzene Adsorbent | |
1 to 14 pH | |
>600 m2/g | |
Use within 5 years from date of manufacture | |
65 to 80% |