missing translation for 'onlineSavingsMsg'
Learn More

Chromogranin A Ab-1 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Chromogranin A Ab-1, Mouse Monoclonal Antibody.

Marca:  Epredia MS-324-P

Dettagli aggiuntivi : Peso : 0.00250kg

 vedi altre versioni di questo prodotto

Codice prodotto. 12671007

  • 501.00€ / 1mL
Spedizione stimata: 20-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Chromogranin A (a protein of 439-amino acids which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.

Host Species: Mouse

Clone: LK2H10

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Pig, Rat and Rabbit. Others not known

Immunogen: Human pheochromocytoma

Molecular Weight: 68-75 kDa

Positive Control: PC-12 cells, Pancreas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
Specifica

Specifica

Chromogranin A Ab-1
Monoclonal
200μg/mL
Mouse
1 mL
Oncogenesis
Human, Non-human Primate, Porcine, Rabbit, Rat
IgG2a κ
Immunohistochemistry (Paraffin), Western Blot
LK2H10
Unconjugated
Human pheochromocytoma
IVD
Primary
Concentrated
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali