Learn More
Chromogranin A Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Chromogranin A Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia MS-324-P0
Dettagli aggiuntivi : Peso : 4.98950kg
Descrizione
Chromogranin A (a protein of 439-amino acids which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.Host Species: Mouse
Clone: LK2H10
Isotype: IgG1/κ
Species Reactivity: Human, Monkey, Pig, Rat and Rabbit. Others not known
Immunogen: Human pheochromocytoma
Molecular Weight: 68-75 kDa
Positive Control: PC-12 cells, Pancreas
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifica
Chromogranin A Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
100 μL | |
Oncogenesis | |
Human, Non-human Primate, Porcine, Rabbit, Rat | |
IgG2a κ |
Immunohistochemistry (Paraffin), Western Blot | |
LK2H10 | |
Unconjugated | |
Human pheochromocytoma | |
IVD | |
Primary | |
Concentrated |