missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHURC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12247-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The MW of this protein is 13 kDa, and 12247-1-AP specifically recognizes the 13 kDa protein.
Transcriptional activator that mediates FGF signaling during neural development. Plays a role in the regulation of cell movement. Does not bind DNA by itself.Specifica
CHURC1 | |
Polyclonal | |
Unconjugated | |
CHURC1 | |
C14orf52, chch, CHURC1, churchill domain containing 1, My015, Protein Churchill | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
211151, 299154, 91612 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6DG52, Q8WUH1 | |
CHURC1 | |
CHURC1 Fusion Protein Ag2892 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |