missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20033-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The antibody recognizes all the isoforms of CIT.
This gene encodes a serine/threonine-protein kinase that functions in cell division. Together with the kinesin KIF14, this protein localizes to the central spindle and midbody, and functions to promote efficient cytokinesis. This protein is involved in central nervous system development. Polymorphisms in this gene are associated with bipolar disorder and risk for schizophrenia. Alternative splicing results in multiple transcript variants.Specifica
CIT | |
Polyclonal | |
Unconjugated | |
CIT | |
CIT, Citron Rho interacting kinase, CRIK, KIAA0949, STK21 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11113, 12704, 83620 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14578, P49025 | |
Cit | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |