missing translation for 'onlineSavingsMsg'
Learn More

CKB Mouse anti-Human, Clone: 2B7C3, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66764-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898935

  • 399.00€ / 150µL
Spedizione stimata: 16-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.
Specifica

Specifica

CKB
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P12277
CKB
CKB Fusion Protein Ag7933
150 μL
Primary
Human
Antibody
IgG2a
Western Blot
2B7C3
Unconjugated
CKB
B CK, CKB, CKBB, Creatine kinase B chain, Creatine kinase B type, creatine kinase, brain
Mouse
Protein A
RUO
1152
-20°C
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali