Learn More
Hirschmann™ Class B Measuring Cylinders with Plastic Socket and Protection Collar, Blue Graduation
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann™ Class B Measuring Cylinders, Blue Graduation, with Plastic Socket and Protection Collar are manufactured from durable DURAN™ glass tubing.
Marca: Hirschmann™ 2260190
Dettagli aggiuntivi : Peso : 0.68338kg
Descrizione
- With spout, plastic socket and protection collar, short line graduation
- Blue graduation
- Class B
- Acc. to DIN EN ISO 4788
- Corrosion-free and resistant to thermal shock
Specifica
Round | |
±3.75 mL | |
B | |
390 mm | |
Vetro (non tipizzato) | |
5 ml |
DIN EN ISO 4788 | |
Hexagonal | |
Graduated Measuring Cylinder | |
Blue | |
500 ml | |
3.75 |