Learn More
Abeba™ Classic 31010 Shoes
Animal fats, oil and fuel resistant. Leather/microfiber construction.
Marca: Abeba™ 31010/42
Dettagli aggiuntivi : Peso : 0.95000kg
Descrizione
- CE EN ISO 20345:2011 SB SRC
- Grained leather black
- Steel toe cap
- Slip resistance PU outsole sole
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Lining microfiber, respiration absorbing
- Adjustable instep area
- Buckle fastening
- Heel patch microfiber
Specifica
CE EN ISO 20345:2011 SB SRC | |
Microfiber | |
42 | |
Polyurethane | |
Steel |
Black | |
Leather | |
Slip-resistant | |
31010 | |
Animal fats, oil and fuel |