Learn More
Buerkle™ Thread adapters LaboPlastBuerkle™ 3/4 in.
StopCock spigots, ball valves and other accessories can be connected to most commercially available containers, with the LaboPlast™ thread adapters.
Marca: Buerkle™ 0600-0094
17.36 EUR valido fino al 2024-12-31
Usa il codice promo "23267" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.04000kg
Descrizione
- All adapters have a standard outer thread R 3/4 in.
Specifica
Thread Adapter | |
Polypropylene, Polyethylene | |
3/4 in. Inflowing, 2 in. Tri-Sure™ outflowing | |
Green/White |